Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HAI-2 Polyclonal Antibody

Rabbit Polyclonal Antibody
$209.50 - $511.50
Specifications
Antigen | HAI-2 |
---|---|
Concentration | 0.5 mg/mL |
Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIPA5119410
![]() |
Invitrogen™
PA5119410 |
200 μL |
Each for $511.50
|
|
|||||
PIPA5121929
|
Invitrogen™
PA5121929 |
20 μL |
Each for $209.50
|
|
|||||
Description
HAI-2 Polyclonal Antibody for Western Blot, IHC (P)
This gene encodes a transmembrane protein with two extracellular Kunitz domains that inhibits a variety of serine proteases. The protein inhibits HGF activator which prevents the formation of active hepatocyte growth factor. This gene is a putative tumor suppressor, and mutations in this gene result in congenital sodium diarrhea. Multiple transcript variants encoding different isoforms have been found for this gene.Specifications
HAI-2 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Polyclonal | |
Liquid | |
IgG | |
Human, Rat | |
SPINT2 | |
AL024025; C76321; DIAR3; FLJ45571; HAI2; HAI-2; Hepatocyte growth factor activator inhibitor type 2; Kop; Kunitz-type protease inhibitor 2; PB; placental bikunin; serine peptidase inhibitor, Kunitz type 2; serine peptidase inhibitor, Kunitz type, 2; serine protease inhibitor, Kunitz type 2; serine protease inhibitor, Kunitz type, 2; Spint2; testicular tissue protein Li 183 | |
SPINT2 | |
Primary | |
Antigen affinity chromatography, Protein A |
0.5 mg/mL | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
RUO | |
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
O43291 | |
10653, 292770 | |
Recombinant protein Serine Peptidase Inhibitor Kunitz Type 2. The antigen corresponds to amino acid range 28-197 of the target protein. | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title