Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HAI-2/SPINT2 Antibody (1), mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB333463 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB333463 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB333463 Supplier Novus Biologicals Supplier No. NBP290373MFV450
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

HAI-2/SPINT2 Monoclonal antibody specifically detects HAI-2/SPINT2 in Mouse samples. It is validated for ELISA

Specifications

Antigen HAI-2/SPINT2
Applications ELISA
Classification Monoclonal
Clone 1
Conjugate mFluor Violet 450 SE
Dilution ELISA
Formulation 50mM Sodium Borate
Gene Alias HAI-2DIAR3, HAI2FLJ45571, Hepatocyte growth factor activator inhibitor type 2, KOP, kunitz-type protease inhibitor 2, PB, Placental bikunin, serine peptidase inhibitor, Kunitz type, 2, serine protease inhibitor, Kunitz type, 2
Host Species Rabbit
Immunogen Rabbit immunized with purified, recombinant Mouse HAI-2/SPINT2 extracellular domain (Accession#: NP_001076017.1, Met 1-Lys 140)
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 10653
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.