Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HAI-2/SPINT2 Antibody (RM0089-7F52), Biotin, Novus Biologicals™
SDP

Catalog No. NBP121728B Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP121728B 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP121728B Supplier Novus Biologicals Supplier No. NBP121728B
Add to Cart
Add to Cart

Rat Monoclonal Antibody

HAI-2/SPINT2 Monoclonal specifically detects HAI-2/SPINT2 in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen HAI-2/SPINT2
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone RM0089-7F52
Conjugate Biotin
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation PBS with 0.05% Sodium Azide
Gene Alias HAI-2DIAR3, HAI2FLJ45571, Hepatocyte growth factor activator inhibitor type 2, KOP, kunitz-type protease inhibitor 2, PB, Placental bikunin, serine peptidase inhibitor, Kunitz type, 2, serine protease inhibitor, Kunitz type, 2
Gene Symbols SPINT2
Host Species Rat
Immunogen Mouse recombinant of the common extracellular region of HAI-2 isoforms
Purification Method Protein A or G purified
Quantity 0.1 mL
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 10653
Test Specificity This antibody detects mouse HIA-2, but not HAI-1 in Western blotting.
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.