Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hairy (Drosophila) Antibody (1/24), Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NP27669A488 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP27669A488 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP27669A488 Supplier Novus Biologicals Supplier No. NBP276659AF488
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Hairy (Drosophila) Monoclonal specifically detects Hairy (Drosophila) in Drosophila samples. It is validated for Western Blot.

Specifications

Antigen Hairy (Drosophila)
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone 1/24
Conjugate Alexa Fluor 488
Dilution Western Blot, Immunohistochemistry
Formulation 50 mM sodium borate with 0.05% sodium azide
Host Species Mouse
Immunogen Hairy gene product
Purification Method Protein A purified
Quantity 0.1 mL
Research Discipline Cellular Signaling, Developmental Biology, Epigenetics, Stem Cells
Primary or Secondary Primary
Test Specificity This antibody will stain Drosophila embryos.
Target Species Drosophila
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.