Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hairy (Drosophila) Antibody (1/24), PerCP, Novus Biologicals™
SDP

Catalog No. NB308504 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB308504 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB308504 Supplier Novus Biologicals Supplier No. NBP276659PCP
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Hairy (Drosophila) Monoclonal antibody specifically detects Hairy (Drosophila) in Drosophila samples. It is validated for Western Blot, Immunohistochemistry, ChIP assay (ChIP)

Specifications

Antigen Hairy (Drosophila)
Applications Western Blot, Immunohistochemistry, ChIP Assay
Classification Monoclonal
Clone 2024-01-24 00:00:00
Conjugate PerCP
Dilution Western Blot, Immunohistochemistry, Chromatin Immunoprecipitation (ChIP)
Formulation PBS
Host Species Mouse
Immunogen Hairy gene product
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Drosophila
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.