Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hairy (Drosophila) Antibody (3/30), DyLight 350, Novus Biologicals™
SDP

Catalog No. NBP250417UV Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP250417UV 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP250417UV Supplier Novus Biologicals Supplier No. NBP250417UV
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Hairy (Drosophila) Monoclonal specifically detects Hairy (Drosophila) in Drosophila samples. It is validated for Western Blot.

Specifications

Antigen Hairy (Drosophila)
Applications Western Blot
Classification Monoclonal
Clone 43189
Conjugate DyLight 350
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Host Species Mouse
Immunogen Hairy gene product
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cellular Signaling, Developmental Biology, Epigenetics, Stem Cells
Primary or Secondary Primary
Target Species Drosophila
Content And Storage Store at 4C in the dark.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.