Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific Supplies Hall Effect Apparatus
SDPSCI_ED

Catalog No. S43710 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Grade:
12 to 15
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S43710 12 to 15
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S43710 Supplier United Scientific Supplies Supplier No. HLEF01
Add to Cart
Add to Cart

The Hall Effect is the generation of a side-to-side voltage in a conductor or semiconductor carrying a current when it is placed in a magnetic field.

  • The Hall Effect is the generation of a side-to-side voltage in a conductor or semiconductor carrying a current when it is placed in a magnetic field.
  • The effect is widely used in magnetic field sensors and is associated with several well-known systematic errors, some of which can be eliminated by measurement techniques.
  • This Hall Effect apparatus consists of a mounted n-type semiconductor chip on a traverse mechanism, a pair of coils, relay-controlled reversing switches, and a control unit with connecting cords.
  • Explores the Hall Effect in a GaAs semiconductor.
  • Explores systematic errors and their elimination. Determines the conductivity of the semiconductor.
  • Uses the semiconductor to plot the coils' magnetic field. n-type GaAs Hall sensor chip.
  • Helmholtz coil pair 75 mm effective diameter, 11.25 mT central magnetic induction at 0.5 A.
  • Two-axis mechanical traverse for field plotting.
  • Constant current sources for magnet (0-0.5 A) and Hall driving current (0—3 mA).
  • Reversing switch for exploring the elimination of systematic error.
  • 3-1/2 digit current and voltage meters.
  • Metal storage case for the Hall Effect board.

Specifications

Grade 12 to 15
Format Kit
Product Type Hall Effect
Languages English
Subject Advanced Physics
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.