Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Halotyrosine Antibody (BTK-94C), Novus Biologicals™
SDP

Catalog No. NB019782 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
100 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB019782 0.025 mg
NBP262808 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB019782 Supplier Novus Biologicals Supplier No. NBP2628080.025MG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Halotyrosine Monoclonal specifically detects Halotyrosine in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunoassay.

Specifications

Antigen Halotyrosine
Applications Western Blot, Immunoprecipitation, In situ PLA, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone BTK-94C
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunoassay
Formulation PBS containing 0.1% BSA with 0.02% Sodium Azide
Host Species Mouse
Immunogen Bromotyrosine structural mimic conjugated to KLH
Purification Method Affinity Purified
Quantity 0.025 mg
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4degreesC.
Isotype IgM
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.