Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Halotyrosine Antibody (BTK-94C), Novus Biologicals™
SDP

Catalog No. NBP262808 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
100 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP262808 100 μg
NB019782 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP262808 Supplier Novus Biologicals Supplier No. NBP262808
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Halotyrosine Monoclonal specifically detects Halotyrosine in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunoassay.

Specifications

Antigen Halotyrosine
Applications Immunocytochemistry, Immunofluorescence, Immunoprecipitation, In Situ Hybridization (ISH), Western Blot
Classification Monoclonal
Clone BTK-94C
Concentration 0.1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunoassay
Formulation PBS containing 0.1% BSA with 0.02% Sodium Azide
Gene Alias bromotyrosine
Host Species Mouse
Immunogen Bromotyrosine structural mimic conjugated to KLH
Purification Method Affinity Purified
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4°C.
Isotype IgM
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.