Learn More
Hamilton™ Tungsten Cleaning Wires
Catalog No. 1481977 Shop All Hamilton Company Products
Click to view available options
For Use With (Equipment):
22, 23G and Larger Needle, Syringe and Needle Cleaning Kit
22s, 25s and 28 to 30G Needle, Syringe and Needle Cleaning Kit
23s Needle, Syringe and Needle Cleaning Kit
24 to 26G Needle, Syringe and Needle Cleaning Kit
26s and 31 to 33G Needle, Syringe and Needle Cleaning Kit
27G Needle, Syringe and Needle Cleaning Kit
For mechanical cleaning to remove needle obstructions.
Tungsten wires, best used with Hamilton Company Needle Cleaning Concentrate (14-819-85, 14-819-86).
Order Info
Ten wires per pack.
Specifications
Material Tungsten
Length (English) 7 in.
Length (Metric) 17.78 cm
Product Type Cleaning Wire
For Use With (Equipment) 27G Needle, Syringe and Needle Cleaning Kit
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.