Learn More
Hamilton™ RN Adapters (Removable Adapters)
Catalog No. p-183827 Shop All Hamilton Company Products
Click to view available options
Type:
Female Luer-lock Hub Adapter
Male Luer-lok Hub Adapter
Adapters provide a connection to threaded fittings, tubing assemblies, tees, and crosses.
Specifications
Type Male Luer-lok Hub Adapter
For Use With Removable Needle Syringe (250μL and Larger)
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.