Learn More
MilliporeSigma™ Supelco™ Hamilton Syringe Cleaner
Shop All MilliporeSigma Supelco ProductsDescription
Cleans Hamilton syringes and other syringes with needles up to 18 gauge (0.049 in. O.D.). Just connect the unit to your water aspirator or other vacuum source, insert the needle through the septum and, in 10-30 seconds, you′re ready for a new sample. A combination of heat (250°C) and vacuum vaporizes and removes contaminating volatiles in a single operation.
No liquids are required with Hamilton Series 7000 syringes. For Series 700 and other conventional syringes, use a rheostat to lower the temperature (to 50°C or below) and maintain a flow of solvent through the barrel and needle.
Instructions are included.
Specifications
Specifications
Material | Metal (Body) |
Description | AC/DC input 120 V AC, L W H 6.0 in. 1.8 in. 2.7 in. |
Quantity | 1/Pk. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.