Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HAND1 Antibody (OTI1G10), DyLight 488, Novus Biologicals™
SDP

Catalog No. NBP271459G Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP271459G 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP271459G Supplier Novus Biologicals Supplier No. NBP271459G
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

HAND1 Monoclonal specifically detects HAND1 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen HAND1
Applications Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI1G10
Conjugate DyLight 488
Dilution Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias BHLHA27, bHLHa27heart- and neural crest derivatives-expressed protein 1, Class A basic helix-loop-helix protein 27, EHAND, Extraembryonic tissues, heart, autonomic nervous system and neural crestderivatives-expressed protein 1, heart and neural crest derivatives expressed 1, Hxt, Thing1
Gene Symbols HAND1
Host Species Mouse
Immunogen Full length recombinant protein of human HAND1 (NP_004812) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 9421.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.