Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Interscience InstaBAG BPW
SDP

Catalog No. 16949061 Shop All Interscience Products
Click to view available options
Capacity (English):
3 oz.
7.6 oz.

The InstaBAG BPW is a filter bag with dehydrated buffered peptone water.

The InstaBAG BPW is a filter bag that includes a dehydrated pre-dosed media, allowing a 2-in-1 process: media and sample preparation for microbiological analyses.

  • Filter bag with dehydrated pre-dosed media
  • Type of dehydrated media: buffered peptone water
  • Filter porosity: <250 microns
  • Compatible with any laboratory blender

Available as 90 mL bag or 225 mL bag.

Specifications

Autoclavable Not Autoclavable
Capacity (English) 7.6 oz.
Capacity (Metric) 225 mL
Color Clear
Disposable Single-use
Sterility Sterile
Thickness (Metric) 72 μm
Latex Free Yes
Thickness (English) 3 mil
Length (English) 7.48 in.
Length (Metric) 190 mm
Width (English) 6.89 in.
Width (Metric) 175 mm
Product Type Blender Bag
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.