Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hantaan Virus Glycoprotein Antibody, Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. NB203284 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB203284 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB203284 Supplier Novus Biologicals Supplier No. NBP241258AF700
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Hantaan Virus Glycoprotein Polyclonal antibody specifically detects Hantaan Virus Glycoprotein in Virus samples. It is validated for ELISA

Specifications

Antigen Hantaan Virus Glycoprotein
Applications ELISA
Classification Polyclonal
Conjugate Alexa Fluor 700
Formulation 50mM Sodium Borate
Gene Alias HTNVsMgp1
Host Species Rabbit
Immunogen Antibody was raised against an 18 amino acid peptide near the amino terminus of the Hantaan virus glycoprotein. The immunogen is located within amino acids 70 - 120 of Hantaan Virus Glycoprotein. Amino Acid Squence: SWRGKADQSQSSQNSFET
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Infections (Virus Bacteria and Parasites)
Primary or Secondary Primary
Gene ID (Entrez) 2943079
Target Species Virus
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.