Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HAP1 Antibody (1B6) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB013831 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB013831 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB013831 Supplier Novus Biologicals Supplier No. NBP280758
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

HAP1 Monoclonal specifically detects HAP1 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen HAP1
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 1B6
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot : 0.5-1.0 ug/ml, Flow Cytometry : 1 ug per million cells, Immunohistochemistry 0.5-1.0 ug/ml, Immunocytochemistry/Immunofluorescence reported in scientific literature, Immunoprecipitation : 1:10-1:500, Immunohistochemistry-Paraffin : 0.5-1.0 ug/ml, Immunohistochemistry-Frozen : 0.5-1.0 ug/ml
Formulation PBS and 50% Glycerol
Gene Alias HAP2huntingtin-associated protein 2, hHLP1, HIP5, HLP1, HLPHAP-1, huntingtin-associated protein 1, Neuroan 1
Gene Symbols HAP1
Host Species Mouse
Immunogen Fusion protein from rat HAP1. [UniProt# P54256]
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Autophagy, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 9001
Test Specificity Human, mouse and rat.
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.