Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HAP1 Antibody (1B6), PE/Cy7, Novus Biologicals™
SDP

Catalog No. N1074569PC7 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N1074569PC7 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N1074569PC7 Supplier Novus Biologicals Supplier No. NB11074569PECY7
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

HAP1 Monoclonal specifically detects HAP1 in Human, Mouse, Rat samples. It is validated for Flow Cytometry.

Specifications

Antigen HAP1
Applications Flow Cytometry
Classification Monoclonal
Clone 1B6
Conjugate PE-Cyanine7
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias HAP2huntingtin-associated protein 2, hHLP1, HIP5, HLP1, HLPHAP-1, huntingtin-associated protein 1, Neuroan 1
Gene Symbols HAP1
Host Species Mouse
Immunogen Fusion protein from rat HAP1. [UniProt No. P54256]
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline Autophagy, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 9001.0
Test Specificity This is specific for isoforms A and B.
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark. Do not freeze.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.