Learn More
hapten 4-hydroxy-3-nitrophenyl acetyl (NP) Antibody (B1-8) - Chimeric - Azide and BSA Free, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | hapten 4-hydroxy-3-nitrophenyl acetyl (NP) |
| Applications | Control, CyTOF, Flow Cytometry, Western Blot |
| Classification | Monoclonal |
| Clone | B1-8 |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:100 - 1:2000, Flow Cytometry 1:10 - 1:1000, CyTOF-ready |
| Formulation | PBS with 0.02% Proclin 300. with 0.02% Proclin 300 |
| Host Species | Rabbit |
| Immunogen | hapten-protein conjugated NP12-chicken gammaglobulin (CG). |
| Purification Method | Protein A or G purified |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.