Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

hapten 4-hydroxy-3-nitrophenyl acetyl (NP) Antibody (B1-8), DyLight 650, Novus Biologicals™
SDP

Catalog No. NB014312 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB014312 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB014312 Supplier Novus Biologicals Supplier No. NBP281027C
Add to Cart
Add to Cart

Human Monoclonal Antibody

hapten 4-hydroxy-3-nitrophenyl acetyl (NP) Monoclonal specifically detects hapten 4-hydroxy-3-nitrophenyl acetyl (NP) in Non-species specific samples. It is validated for Western Blot, Flow Cytometry.

Specifications

Antigen hapten 4-hydroxy-3-nitrophenyl acetyl (NP)
Applications Western Blot, Flow Cytometry
Classification Monoclonal
Clone B1-8
Conjugate DyLight 650
Dilution Western Blot, Flow Cytometry
Formulation 50mM Sodium Borate
Gene Alias (4-Hydroxy-3-nitrophenyl)acetyl, 2-(4-Hydroxy-3-nitrophenyl)acetate, 2-(4-Hydroxy-3-nitrophenyl)acetic acid, 3-nitro-4-Hydroxyphenylacetic acid, 4-Hydroxy-3-nitrophenyl acetyl
Host Species Human
Immunogen hapten-protein conjugated NP12-chicken gammaglobulin (CG).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Hapten (4-hydroxy-3-nitropheny1) acetyl (NP).
Target Species Non-species specific
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 λ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.