Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
hapten 4-hydroxy-3-nitrophenyl acetyl (NP) Antibody (B1-8) - Chimeric - Azide and BSA Free, Novus Biologicals™

Rabbit Monoclonal Antibody
$268.00 - $584.50
Specifications
| Antigen | hapten 4-hydroxy-3-nitrophenyl acetyl (NP) |
|---|---|
| Clone | B1-8 |
| Dilution | Western Blot 1:100 - 1:2000, Flow Cytometry 1:10 - 1:1000, CyTOF-ready |
| Applications | Western Blot, Flow Cytometry, CyTOF |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP2526425
![]() |
Novus Biologicals
NBP2526400.025MG |
0.025 mg |
Each for $268.00
|
|
|||||
NBP25264002
![]() |
Novus Biologicals
NBP2526400.2MG |
0.2 mg |
Each for $584.50
|
|
|||||
Description
hapten 4-hydroxy-3-nitrophenyl acetyl (NP) Monoclonal specifically detects hapten 4-hydroxy-3-nitrophenyl acetyl (NP) in Non-species specific samples. It is validated for Western Blot, Flow Cytometry, Control, CyTOF-ready.Specifications
| hapten 4-hydroxy-3-nitrophenyl acetyl (NP) | |
| Western Blot 1:100 - 1:2000, Flow Cytometry 1:10 - 1:1000, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS with 0.02% Proclin 300. with 0.02% Proclin 300 | |
| IgG | |
| Protein A or G purified | |
| Hapten (4-hydroxy-3-nitropheny1) acetyl (NP). |
| B1-8 | |
| Western Blot, Flow Cytometry, CyTOF | |
| Unconjugated | |
| Rabbit | |
| Non-species specific | |
| hapten-protein conjugated NP12-chicken gammaglobulin (CG). | |
| Primary | |
| Store at 4C for up to 3 months. For longer storage, aliquot and store at -20C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title