Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HARBI1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $487.50
Specifications
Antigen | HARBI1 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP17423020
![]() |
Novus Biologicals
NBP17423020UL |
20 μL |
Each for $206.00
|
|
|||||
NBP174230
![]() |
Novus Biologicals
NBP174230 |
100 μL |
Each for $487.50
|
|
|||||
Description
HARBI1 Polyclonal specifically detects HARBI1 in Mouse samples. It is validated for Western Blot.Specifications
HARBI1 | |
Polyclonal | |
Rabbit | |
C11orf77, chromosome 11 open reading frame 77, EC 3.1, FLJ32675, harbinger transposase derived 1, Harbinger transposase-derived nuclease, putative nuclease HARBI1 | |
HARBI1 | |
IgG | |
27 kDa |
Western Blot | |
Unconjugated | |
RUO | |
283254 | |
Synthetic peptides corresponding to the N terminal of Harbi1. Immunizing peptide sequence ITVLDCDLLLYGRGHRTLDRFKLDDVTDEYLMSMYGFPRQFIYFLVELLG. | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title