Learn More
HAUS3 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A305738AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
Contributes to mitotic spindle assembly, maintece of centrosome integrity and completion of cytokinesis as part of the HAUS augmin-like complex.Specifications
HAUS3 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q68CZ6 | |
HAUS3 | |
Between 75 and 125 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunoprecipitation | |
Unconjugated | |
HAUS3 | |
HAUS augmin-like complex subunit 3, HAUS3, C4orf15 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
79441 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.