Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HAUS3 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$151.31
Specifications
Antigen | HAUS3 |
---|---|
Applications | Immunoprecipitation |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-157-1121
![]() |
Bethyl Laboratories, Inc
A305738AT |
20 μL |
Each of 1 for $151.31
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt.
Contributes to mitotic spindle assembly, maintece of centrosome integrity and completion of cytokinesis as part of the HAUS augmin-like complex.Specifications
HAUS3 | |
Polyclonal | |
Liquid | |
RUO | |
TBS with 0.1% BSA and 0.09% sodium azide | |
HAUS augmin-like complex subunit 3, HAUS3, C4orf15 | |
HAUS3 | |
IgG | |
Antigen affinity chromatography | |
Antibody |
Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Human | |
Q68CZ6 | |
79441 | |
Between 75 and 125 | |
Primary | |
4° C | |
HAUS3 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title