Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HAUS8, p Ser70 Antibody, Novus Biologicals™
SDP

Catalog No. NBP219128 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP219128 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP219128 Supplier Novus Biologicals Supplier No. NBP219128
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

HAUS8 Polyclonal specifically detects HAUS8 in Human samples. It is validated for Western Blot, ELISA.

Specifications

Antigen HAUS8 (p Ser70)
Applications Western Blot, ELISA
Classification Polyclonal
Concentration 1.22 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1.22 ug/ml, ELISA 1:17000-1:45000
Formulation 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide
Gene Accession No. Q9BT25
Gene Alias DGT4, HAUS Augmin Like Complex Subunit 8, HEC1/NDC80 interacting, centrosome associated 1, HEC1/NDC80-Interacting Centrosome-Associated Protein 1, Hec1-Interacting And Centrosome-Associated 1, HICE1, NY-SAR-48, Sarcoma Antigen NY-SAR-48
Gene Symbols HAUS8
Host Species Rabbit
Immunogen HAUS8 [p Ser70] Antibody was prepared by repeated immunizations with a phosphorylated synthetic peptide corresponding to the region of amino acids containing serine 70 of HAUS8. (Uniprot: Q9BT25)
Molecular Weight of Antigen 44.86 kDa
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 93323
Test Specificity HAUS8 pS70 affinity purified antibody is directed against the phosphorylated form of human HAUS8 protein at the S70 residue. The resultant affinity purified antibody was then cross adsorbed against the non-phosphorylated form of the immunizing peptide. The antibody is specific for the phosphorylated form of HAUS8. Reactivity with non-phosphorylated human HAUS8 is minimal by ELISA and western blot. A BLAST analysis was used to suggest cross reactivity with HAUS8 from human based on 100% sequence homology with the immunogen. Reactivity against homologues from other sources is not known.
Target Species Human
Content And Storage Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.