Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HAX-1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
| Antigen | HAX-1 |
|---|---|
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Regulatory Status | RUO |
Description
HAX-1 Polyclonal specifically detects HAX-1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin).Specifications
| HAX-1 | |
| Unconjugated | |
| RUO | |
| FLJ17042, FLJ93803, HAX-1, HCLS1 (and PKD2) associated protein, HCLS1 associated protein X-1, HCLS1-associated protein X-1, HCLSBP1, HS1 binding protein, HS1-associating protein X-1, HS1-binding protein 1, HS1BP1FLJ18492, HSP1BP-1, SCN3 | |
| HAX1 | |
| IgG | |
| 31 kDa |
| Polyclonal | |
| Rabbit | |
| O00165 | |
| 10456 | |
| Synthetic peptides corresponding to HAX1(HCLS1 associated protein X-1) The peptide sequence was selected from the middle region of HAX1. Peptide sequence LPGPESETPGERLREGQTLRDSMLKYPDSHQPRIFGGVLESDARSESPQP. | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title