Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ HaXS8
GREENER_CHOICE

Catalog No. 49912
Click to view available options
Quantity:
2 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical dimerizer

HaXS8 is a promotes covalent and irreversible HaloTag™ and SNAP-tag™ substrate dimerization. Cell permeable.

HaloTag is a trademark of Promega Corporation, and SNAP-tag is a trademark of New England BioLabs, Inc.

Specifications

Chemical Name or Material N-[[4-[[(2-Amino-9-H-purin-6-yl)oxy]methyl]phenyl]methyl]-2-[4-[(18-chloro-3,6,9,12-tetraoxaoctadec-1yl)oxy]-2,3,5,6-tetrafluorophenoxy]acetamide
CAS 2080306-25-6
Quantity 2 mg
Solubility Soluble to 100mM in DMSO
Molecular Formula C35H43ClF4N6O8
Formula Weight 787.2
Product Type Chelator

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.