Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HB-EGF Antibody (125923), Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NB364687 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB364687 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB364687 Supplier Novus Biologicals Supplier No. FAB259X
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

HB-EGF Monoclonal antibody specifically detects HB-EGF in Human samples. It is validated for Western Blot

Specifications

Antigen HB-EGF
Applications Western Blot
Classification Monoclonal
Clone 125923
Conjugate Alexa Fluor 532
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias diphtheria toxin receptor (heparin-binding epidermal growth factor-like growthfactor), DTRHEGFLdiphtheria toxin receptor (heparin-binding EGF-like growth factor), DTS, DTSF, heparin-binding EGF-like growth factor, heparin-binding epidermal growth factor, proheparin-binding EGF-like growth factor
Host Species Mouse
Immunogen S. frugiperda insect ovarian cell line Sf 21-derived recombinant human HB-EGF, Asp63-Leu148, Accession # Q53H93
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins
Primary or Secondary Primary
Gene ID (Entrez) 1839
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.