Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HBcAg Antibody (H3A4cc), DyLight 550, Novus Biologicals™
SDP

Catalog No. NB1102512R Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1102512R 0.2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1102512R Supplier Novus Biologicals Supplier No. NB1102512R
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

HBcAg Monoclonal specifically detects HBcAg in Virus samples. It is validated for Western Blot, ELISA.

Specifications

Antigen HBcAg
Applications Western Blot, ELISA
Classification Monoclonal
Clone H3A4cc
Conjugate DyLight 550
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias c, Core antigen, HBc, HBcAg, Hepatitis B Virus core antigen
Gene Symbols C
Host Species Mouse
Immunogen Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with recombinant HBcAg.
Purification Method Protein G purified
Quantity 0.2 mL
Regulatory Status RUO
Research Discipline Virology Bacteria and Parasites
Primary or Secondary Primary
Gene ID (Entrez) 944568
Test Specificity Hepatitis B Core Antigen (H3A4)
Target Species Virus
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.