Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HBcAg Antibody (Hep2 (H69)), Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB10064454X Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10064454X 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10064454X Supplier Novus Biologicals Supplier No. NB10064454AF488
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

HBcAg Monoclonal specifically detects HBcAg in Virus samples. It is validated for Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin).

Specifications

Antigen HBcAg
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone Hep2 (H69)
Conjugate Alexa Fluor 488
Dilution Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias c, Core antigen, HBc, HBcAg, Hepatitis B Virus core antigen
Gene Symbols C
Host Species Mouse
Immunogen Purified hepatitis B core antigen (HBcAg )
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Virology Bacteria and Parasites
Primary or Secondary Primary
Gene ID (Entrez) 944568
Test Specificity NB100-64454 is specific for Hepatitis B core antigen (HBcAg).
Target Species Virus
Content And Storage Store at 4C in the dark.
Isotype IgG3
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.