Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HBG1 Rabbit anti-Human, Clone: 1R9T7, Novus Biologicals™
SDP

Catalog No. NB165692 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB165692 100 μg
NB165693 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB165692 Supplier Novus Biologicals Supplier No. NBP315298100UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

HBG1 Monoclonal antibody specifically detects HBG1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence

Specifications

Antigen HBG1
Applications Western Blot, Immunohistochemistry, Immunofluorescence
Classification Monoclonal
Clone 1R9T7
Conjugate Unconjugated
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:50 - 1:200
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias gamma-1-globin, Hb F Agamma, HBGA, HBGR, HBG-T2, Hemoglobin gamma A, Hemoglobin gamma-1 chain, Hemoglobin gamma-A chain, Hemoglobin subunit gamma 1, HSGGL1, PRO2979
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 68-147 of human HBG1 (P69891).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3047
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.