Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HBLD1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP15471820 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP15471820 20 μL
NBP154718 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP15471820 Supplier Novus Biologicals Supplier No. NBP15471820UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

HBLD1 Polyclonal specifically detects HBLD1 in Human samples. It is validated for Western Blot.

Specifications

Antigen HBLD1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000
Formulation PBS and 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. Q86U28
Gene Alias c14_5557, HBLD1, HesB like domain containing 1, HESB-like domain-containing protein 1, iron-sulfur cluster assembly 2 homolog (S. cerevisiae), iron-sulfur cluster assembly 2 homolog, mitochondrial, ISA2
Gene Symbols ISCA2
Host Species Rabbit
Immunogen Synthetic peptides corresponding to ISCA2(iron-sulfur cluster assembly 2 homolog (S. cerevisiae)) The peptide sequence was selected from the middle region of ISCA2. Peptide sequence RREASSSSPEAGEGQIRLTDSCVQRLLEITEGSEFLRLQVEGGGCSGFQY.
Molecular Weight of Antigen 16 kDa
Purification Method Affinity Purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 122961
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.