Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HBP1 Antibody, PE, Novus Biologicals™
SDP

Catalog No. NB351503 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB351503 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB351503 Supplier Novus Biologicals Supplier No. NBP306527PE
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

HBP1 Polyclonal antibody specifically detects HBP1 in Human samples. It is validated for ELISA

Specifications

Antigen HBP1
Applications ELISA
Classification Polyclonal
Conjugate PE
Dilution ELISA
Formulation PBS
Gene Alias FLJ16340, High mobility group box transcription factor 1, HMG box transcription factor 1, HMG box-containing protein 1, HMG-box containing protein 1, HMG-box transcription factor 1, USMCC2
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Human HBP1 (Accession#: O60381-1; Pro208-Phe345)
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cardiovascular Biology, Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 26959
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.