Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HBS1L Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
| Antigen | HBS1L |
|---|---|
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Regulatory Status | RUO |
Description
HBS1L Polyclonal specifically detects HBS1L in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunofluorescence.Specifications
| HBS1L | |
| Unconjugated | |
| RUO | |
| Q9Y450 | |
| 10767 | |
| Synthetic peptides corresponding to HBS1L(HBS1-like (S. cerevisiae)) The peptide sequence was selected from the C terminal of HBS1L. Peptide sequence MNHKILVCFADQGSGFCITGKIEAGYIQTGDRLLAMPPNETCTVKGITLH. | |
| Primary |
| Polyclonal | |
| Rabbit | |
| Signal Transduction | |
| DKFZp434g247, ERF3-similar protein, ERFSDKFZp686L13262, HBS1 (S. cerevisiae)-like, HBS1KIAA1038EF-1a, HBS1-like (S. cerevisiae), HBS1-like protein, Hsp70 subfamily B suppressor 1-like protein, HSPC276 | |
| HBS1L | |
| IgG | |
| 22 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title