Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HBS1L Antibody (OTI1C1), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB288747 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB288747 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB288747 Supplier Novus Biologicals Supplier No. NBP271628APC
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

HBS1L Monoclonal antibody specifically detects HBS1L in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen HBS1L
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone OTI1C1
Conjugate APC
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias DKFZp434g247, ERF3-similar protein, ERFSDKFZp686L13262, HBS1 (S. cerevisiae)-like, HBS1KIAA1038EF-1a, HBS1-like (S. cerevisiae), HBS1-like protein, Hsp70 subfamily B suppressor 1-like protein, HSPC276
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 1-260 of human HBS1L (NP_006611) produced in E.coli.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 10767
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.