Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HBS1L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556241
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556241 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556241 Supplier Invitrogen™ Supplier No. PA556241
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RNVRGYNYDE DFEDDDLYGQ SVEDDYCISP STAAQFIYSR RDKPSVEPVE EYDYEDLKES SNSVSNHQLS GFDQARLYSC LDHMREVLGD AVPDEI Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 88%.

This gene encodes a member of the GTP-binding elongation factor family. It is expressed in multiple tissues with the highest expression in heart and skeletal muscle. The intergenic region of this gene and the MYB gene has been identified to be a quantitative trait locus (QTL) controlling fetal hemoglobin level, and this region influnces erythrocyte, platelet, and monocyte counts as well as erythrocyte volume and hemoglobin content. DNA polymorphisms at this region associate with fetal hemoglobin levels and pain crises in sickle cell disease. A single nucleotide polymorphism in exon 1 of this gene is significantly associated with severity in beta-thalassemia/Hemoglobin E. Multiple alternatively spliced transcript variants encoding different protein isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HBS1L
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene HBS1L
Gene Accession No. Q9Y450
Gene Alias 2810035F15Rik; AI326327; EF-1a; eRF3 family member; eRF3c; ERF3-similar protein; ERFS; eukaryotic release factor 3; Hbs1; HBS1 like translational GTPase; HBS1L; Hbs1-like (S. cerevisiae); HBS1-like protein; HBS1-like translational GTPase; Hsp70 subfamily B suppressor 1-like protein; HSPC276; Kiaa1038
Gene Symbols HBS1L
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human HBS1L. Recombinant protein control fragment (Product #RP-94690).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10767
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.