Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HBsAg Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB10062647 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
0.5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10062647 0.5 mL
NB017502 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10062647 Supplier Novus Biologicals Supplier No. NB10062647
Add to Cart
Add to Cart

Goat Polyclonal Antibody

HBsAg Polyclonal specifically detects HBsAg in Virus samples. It is validated for ELISA.

Specifications

Antigen HBsAg
Applications ELISA
Classification Polyclonal
Concentration 4.0 mg/mL
Conjugate Unconjugated
Dilution ELISA 1:200 - 1:1000
Formulation Phosphate Buffered Saline with 0.1% Sodium Azide
Gene Alias HBsAg, HBV major surface antigen, HBV surface antigen, Hepatitis B Virus major surface antigen, Hepatitis B Virus Surface Antigen, Major surface antigen, protein S
Gene Symbols S
Host Species Goat
Immunogen Hepatitis B, subtypes ad and ay
Purification Method IgG purified
Quantity 0.5 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 944569
Test Specificity NB100-62647 is specific for Hepatitis B surface antigen (HBsAg), a protein component of the viral envelope which is predominantly expressed in the cytoplasm of infected cells. Hep B is a major causative agent of acute and chronic liver disease in humans.
Target Species Virus
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.