Learn More
Description
Specifications
Specifications
| Antigen | HBsAg |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | HB11 |
| Conjugate | Alexa Fluor 647 |
| Dilution | ELISA, Sandwich ELISA |
| Formulation | 50mM Sodium Borate with 0.05% Sodium Azide |
| Gene Alias | HBsAg, HBV major surface antigen, HBV surface antigen, Hepatitis B Virus major surface antigen, Hepatitis B Virus Surface Antigen, Major surface antigen, protein S |
| Gene Symbols | S |
| Host Species | Mouse |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
