Learn More
Description
Specifications
Specifications
| Antigen | HBsAg |
| Applications | ELISA, Immunocytochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Clone | HB3 |
| Conjugate | DyLight 488 |
| Dilution | ELISA, Immunocytochemistry/Immunofluorescence |
| Gene Alias | HBsAg, HBV major surface antigen, HBV surface antigen, Hepatitis B Virus major surface antigen, Hepatitis B Virus Surface Antigen, Major surface antigen, protein S |
| Gene Symbols | S |
| Host Species | Mouse |
| Immunogen | Purified HBsAg from human plasma. |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
