Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HBsAg Antibody (HB3), DyLight 550, Novus Biologicals™
SDP

Catalog No. NB500474R Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB500474R 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB500474R Supplier Novus Biologicals Supplier No. NB500474R
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

HBsAg Monoclonal specifically detects HBsAg in Virus samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen HBsAg
Applications ELISA, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone HB3
Conjugate DyLight 550
Dilution ELISA, Immunocytochemistry/Immunofluorescence
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias HBsAg, HBV major surface antigen, HBV surface antigen, Hepatitis B Virus major surface antigen, Hepatitis B Virus Surface Antigen, Major surface antigen, protein S
Gene Symbols S
Host Species Mouse
Immunogen Purified HBsAg from human plasma.
Purification Method Ion exchange chromatography
Quantity 0.1 mL
Research Discipline Cell Biology, Virology Bacteria and Parasites
Primary or Secondary Primary
Gene ID (Entrez) 944569.0
Test Specificity The antibody HB3 recognizes following Hepatitis B virus (HBV) subtypes: ayw2, ayw3, ayw4, ayr, adw2, adw4, adrq+, adrq-. Hepatitis B surface antigen (HBsAg) is a marker of infectivity. Specificity of the antibody HB3 was verified by ELISA on panel of virus subtypes identified on International Workshop on HBsAg Subtypes (Paris, April 1975). The antibody HB3 does not cross-block with the antibody HB5.
Target Species Virus
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.