Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HBsAg Antibody (HB3), FITC, Novus Biologicals™
SDP

Catalog No. NB359251 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB359251 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB359251 Supplier Novus Biologicals Supplier No. NB500474F
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

HBsAg Monoclonal antibody specifically detects HBsAg in Virus samples. It is validated for ELISA, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen HBsAg
Applications ELISA, Immunocytochemistry
Classification Monoclonal
Clone HB3
Conjugate FITC
Dilution ELISA, Immunocytochemistry/ Immunofluorescence
Formulation PBS
Gene Alias HBsAg, HBV major surface antigen, HBV surface antigen, Hepatitis B Virus major surface antigen, Hepatitis B Virus Surface Antigen, Major surface antigen, protein S
Host Species Mouse
Immunogen Purified HBsAg from human plasma.
Purification Method Ion exchange chromatography
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Virology Bacteria and Parasites
Primary or Secondary Primary
Gene ID (Entrez) 944569
Target Species Virus
Content And Storage Store at 4C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.