Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ HC 030031
GSA_VAGREENER_CHOICESDP

Catalog No. 289610 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
28-961-0 10 mg
28-965-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 28-961-0 Supplier Tocris Bioscience™ Supplier No. 2896/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Selective TRPA1 blocker

Selective TRPA1 channel blocker that antagonizes AITC- and formalin-evoked calcium influx (IC50 values are 6.2 and 5.3 μM respectively). Does not block currents mediated by TRPV1, TRPV3, TRPV4, hERG or NaV1.2 channels. Inhibits AITC- and formalin-induced flinching in vivo.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Chemical Identifiers

CAS 349085-38-7
Molecular Formula C18H21N5O3

Specifications

Quantity 10 mg
Formula Weight 355.39
Purity >99%
Molecular Weight (g/mol) 355.398
Chemical Name or Material HC 030031
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.