Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HCFC1R1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP231788 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP231788 0.1 mL
NB437196 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP231788 Supplier Novus Biologicals Supplier No. NBP231788
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

HCFC1R1 Polyclonal specifically detects HCFC1R1 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin).

Specifications

Antigen HCFC1R1
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:20 - 1:50
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Accession No. Q9NWW0
Gene Alias FLJ20568, HCF-1 beta-propeller interacting protein, HCF-1 beta-propeller-interacting protein, host cell factor C1 regulator 1, host cell factor C1 regulator 1 (XPO1 dependant), host cell factor C1 regulator 1 (XPO1 dependent), HPIPMGC99622, MGC70711
Gene Symbols HCFC1R1
Host Species Rabbit
Immunogen This antibody was developed against a recombinant protein corresponding to amino acids: QPLQRGPQGGAQRLPRAALGVTWGLDASSPLRGAVPMSTKRRLEEEQEPLRKQFLSEEN
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54985
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.