Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

hCG Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB120185 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB120185 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB120185 Supplier Novus Biologicals Supplier No. NBP311024
Add to Cart
Add to Cart

Goat Polyclonal Antibody

hCG Polyclonal specifically detects hCG in Human samples. It is validated for Western Blot, ELISA.

Specifications

Antigen hCG
Applications Western Blot, ELISA
Classification Polyclonal
Concentration 4.5 mg/ml
Conjugate Unconjugated
Dilution Western Blot, ELISA
Formulation 10 mM Sodium Phosphate, 0.15 M Sodium Chloride, pH 7
Host Species Goat
Immunogen Human TSH (intact)
Purification Method >95%, by SDS-PAGE
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4°C.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.