Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
hCG beta 3 Mouse anti-Human, Unconjugated, Clone: P3W80, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MABS206225UL
Description
hCG beta 3 Monoclonal antibody specifically detects hCG beta 3 in Human samples. It is validated for ELISA, Flow Cytometry, Immunohistochemistry (Paraffin).
Specifications
hCG beta 3 | |
Monoclonal | |
Unconjugated | |
Choriogonadotropin subunit beta 3;Choriogonadotropin subunit beta;CG-beta;Chorionic gonadotropin chain beta | |
Mouse | |
17.74 kDa calculated. | |
25 μL | |
Primary | |
Human | |
Purified |
ELISA, Flow Cytometry, Immunohistochemistry (Paraffin) | |
P3W80 | |
NP_000728 | |
CGB3, CGB, CGB5, CBG8 | |
Purified human chorionic gonadotropin, beta subunit. | |
Protein G Purified | |
Signaling | |
Clone P3W80 is a mouse monoclonal antibody that detects human Choriogonadotropin subunit beta 3. | |
Stable for 1 year at -20°C from date of receipt. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction