Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
hCG beta 3 Mouse anti-Human, Unconjugated, Clone: P3W80, MilliporeSigma™
Mouse Monoclonal Antibody
$182.31 - $457.74
Specifications
Antigen | hCG beta 3 |
---|---|
Clone | P3W80 |
Applications | ELISA, Flow Cytometry, Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MABS20622MI
|
MilliporeSigma
MABS206225UL |
25 μL |
Each for $182.31
|
|
|||||
MABS20621MI
|
MilliporeSigma
MABS2062100UL |
100 μL |
Each for $457.74
|
|
|||||
Description
hCG beta 3 Monoclonal antibody specifically detects hCG beta 3 in Human samples. It is validated for ELISA, Flow Cytometry, Immunohistochemistry (Paraffin).Specifications
hCG beta 3 | |
ELISA, Flow Cytometry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Human | |
Choriogonadotropin subunit beta 3;Choriogonadotropin subunit beta;CG-beta;Chorionic gonadotropin chain beta | |
Purified human chorionic gonadotropin, beta subunit. | |
Primary | |
Stable for 1 year at -20°C from date of receipt. | |
17.74 kDa calculated. |
P3W80 | |
Monoclonal | |
Purified | |
Signaling | |
NP_000728 | |
CGB3, CGB, CGB5, CBG8 | |
IgG1 κ | |
Protein G Purified | |
Clone P3W80 is a mouse monoclonal antibody that detects human Choriogonadotropin subunit beta 3. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title