Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HCN3 Antibody, Novus Biologicals™
SDP

Catalog No. NBP168869 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP168869 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP168869 Supplier Novus Biologicals Supplier No. NBP168869
Add to Cart
Add to Cart

Goat Polyclonal Antibody

HCN3 Polyclonal specifically detects HCN3 in Rat samples. It is validated for Western Blot, Peptide ELISA.

Specifications

Antigen HCN3
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1 - 3 ug/ml, Peptide ELISA Detection limit 1:32000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_065948.1
Gene Alias hyperpolarization activated cyclic nucleotide-gated potassium channel 3, KIAA1535MGC131493, potassium/sodium hyperpolarization-activated cyclic nucleotide-gated channel 3
Gene Symbols HCN3
Host Species Goat
Immunogen Peptide with sequence C-RGVRGRAPSTGAQ corresponding to internal region according to NP_065948.1.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 57657
Test Specificity Cross reacts with Rat. Expected from sequence similarity: Human, Mouse, Dog and Cow.
Target Species Rat
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.