Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HCN4 Antibody (S114/10), DyLight 405, Novus Biologicals™
SDP

Catalog No. NBP212910V Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP212910V 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP212910V Supplier Novus Biologicals Supplier No. NBP212910V
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

HCN4 Monoclonal specifically detects HCN4 in Human, Mouse, Rat, Rabbit samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen HCN4
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Monoclonal
Clone S114/10
Conjugate DyLight 405
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Gene Alias hyperpolarization activated cyclic nucleotide-gated cation channel 4, hyperpolarization activated cyclic nucleotide-gated potassium channel 4, potassium/sodium hyperpolarization-activated cyclic nucleotide-gated channel 4, SSS2
Gene Symbols HCN4
Host Species Mouse
Immunogen Fusion protein amino acids 1-019-1198 (c-terminus) of rat HCN4
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10021
Test Specificity Detects approx 130 kDa. No cross-reactivity against other HCNs.
Target Species Human, Mouse, Rat, Rabbit
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.