Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HCP1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP106603 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP106603 0.2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP106603 Supplier Novus Biologicals Supplier No. NBP106603
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

HCP1 Polyclonal specifically detects HCP1 in Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen HCP1
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1:100-1:1000, ELISA 1:100-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50 - 1:200, Immunohistochemistry-Paraffin
Formulation Antiserum with No Preservative
Gene Alias FLJ39875, G21, HCP1PCFT/HCP1, Heme carrier protein 1, MGC9564, PCFTsolute carrier family 46, member 1, proton-coupled folate transporter, solute carrier family 46 (folate transporter), member 1, Solute carrier family 46 member 1
Gene Symbols SLC46A1
Host Species Rabbit
Immunogen Synthetic peptide derived from Cter domain of mouse HCP1 protein
Purification Method Unpurified
Quantity 0.2 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 113235
Test Specificity Reacts specifically with mouse 50 kDa protein
Reconstitution Reconstitute with 0.2 ml distilled water.
Target Species Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.