Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HCV E1E2 Human anti-Virus, Clone: AR5A, MilliporeSigma™
Human Monoclonal Antibody
$182.31 - $468.00
Specifications
Antigen | HCV E1E2 |
---|---|
Clone | AR5A |
Concentration | 0.5 mg/mL |
Applications | ELISA, Immunoprecipitation, Neutralization |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MABF233425
|
MilliporeSigma
MABF233425UL |
25 μL |
Each for $182.31
|
|
|||||
MABF2334100
|
MilliporeSigma
MABF2334100UL |
100 μL |
Each for $468.00
|
|
|||||
Description
HCV E1E2 Monoclonal antibody specifically detects HCV E1E2 in Virus samples. It is validated for ELISA, Immunoprecipitation, Neutralizing.Specifications
HCV E1E2 | |
0.5 mg/mL | |
Monoclonal | |
Purified | |
RUO | |
Purified human recombinant monoclonal antibody in PBS without preservatives. | |
IgG1 | |
Protein G purified |
AR5A | |
ELISA, Immunoprecipitation, Neutralization | |
Unconjugated | |
Human | |
Virus | |
Antibody Fab fragment V1 expressed in E. coli, purified, and converted into full-length IgG1 molecules in CHO-K1 cells. | |
Primary | |
Stable for 1 year at -10°C to -25°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title