Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HCV E2 Mouse anti-Hepatitis C virus, Unconjugated, Clone: 19B3, MilliporeSigma™
Mouse Monoclonal Antibody
$184.08 - $457.74
Specifications
Antigen | HCV E2 |
---|---|
Clone | 19B3 |
Applications | ELISA, Neutralization, Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MABF23082MI
|
MilliporeSigma
MABF230825UG |
25 μg |
Each for $184.08
|
|
|||||
MABF23081MI
|
MilliporeSigma
MABF2308100UG |
100 μg |
Each for $457.74
|
|
|||||
Description
HCV E2 Monoclonal antibody specifically detects HCV E2 in Hepatitis C virus samples. It is validated for ELISA, Neutralizing, Western Blotting.Specifications
HCV E2 | |
ELISA, Neutralization, Western Blot | |
Unconjugated | |
Mouse | |
Hepatitis C virus | |
DNA constructs of E1E2 followed by a boost with truncated E2 protein (aa 412-645). | |
Primary | |
Stable for 1 year at -20°C from date of receipt. | |
327.15 kDa calculated for the entire genome polyprotein. |
19B3 | |
Monoclonal | |
Purified | |
Inflammation and Immunology | |
Hepatittis C virus E2 protein | |
IgG2a κ | |
Protein G Purified | |
Clone 19B3 is a mouse monoclonal antibody that detects the envelope glycoprotein E2 of the Hepatitis C virus. It targets an epitope within 12 amino acids from the N-terminal region of the E2. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title